Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCE Antibody (L5-P1E6*G5), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP25040A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP25040A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP25040A532 Supplier Novus Biologicals Supplier No. NBP250400AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FANCE Monoclonal specifically detects FANCE in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen FANCE
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone L5-P1E6*G5
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FACEFAE, Fanconi anemia group E protein, Fanconi anemia, complementation group E, Protein FACE
Gene Symbols FANCE
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide - KAALKHLGP
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2178
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.