Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCG Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB052558 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052558 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052558 Supplier Novus Biologicals Supplier No. NBP298109G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FANCG Polyclonal specifically detects FANCG in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FANCG
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DNA repair protein XRCC9, FAG, Fanconi anemia group G protein, Fanconi anemia, complementation group G, Protein FACG, X-ray repair, complementing defective, in Chinese hamster, 9, XRCC9X-ray repair complementing defective repair in Chinese hamster cells 9
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FANCG fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2189
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.