Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCM Antibody (CV5.1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP250418AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250418AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250418AH Supplier Novus Biologicals Supplier No. NBP250418AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FANCM Monoclonal specifically detects FANCM in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FANCM
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CV5.1
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ATP-dependent RNA helicase FANCM, EC 3.6.1, FAAP250EC 3.6.4.13, Fanconi anemia, complementation group M, Fanconi anemia-associated polypeptide of 250 kDa, KIAA1596Fanconi anemia group M protein, MGC176453, Protein FACM, Protein Hef ortholog
Gene Symbols FANCM
Host Species Mouse
Immunogen His-tagged denatured FANCM (aa 1507-1679) made in E.coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 57697
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.