Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAPP2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB136201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB136201 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB136201 Supplier Novus Biologicals Supplier No. NBP306343AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAPP2 Polyclonal specifically detects FAPP2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FAPP2
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZp313M1034, DKFZp686J09164, FAPP-2, FAPP2MGC3358, hFAPP2, PH domain-containing family A member 8, Phosphatidylinositol-four-phosphate adapter protein 2, Phosphoinositol 4-phosphate adapter protein 2, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 8, pleckstrin homology domain-containing family A member 8, Serologically defined breast cancer antigen NY-BR-86
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FAPP2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84725
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.