Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fargesin 98.0+%, TCI America™
SDP

Catalog No. F118810MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
10 mg
50 mg

Chemical Identifiers

CAS 31008-19-2
Molecular Formula C21H22O6
Molecular Weight (g/mol) 370.40
MDL Number MFCD07781423
InChI Key AWOGQCSIVCQXBT-UHFFFAOYNA-N
Synonym rel-5-[(1R,3aS,4S,6aS)-4-(3,4-Dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl]-1,3-benzodioxole
PubChem CID 92966415
IUPAC Name 5-[4-(3,4-dimethoxyphenyl)-hexahydrofuro[3,4-c]furan-1-yl]-2H-1,3-benzodioxole
SMILES COC1=C(OC)C=C(C=C1)C1OCC2C1COC2C1=CC2=C(OCO2)C=C1

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 10 mg
Formula Weight 370.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Fargesin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.