Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FARSB Antibody (OTI4B3), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288802 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288802 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288802 Supplier Novus Biologicals Supplier No. NBP271539APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FARSB Monoclonal antibody specifically detects FARSB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen FARSB
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4B3
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias EC 6.1.1, EC 6.1.1.20, FARSLBphenylalanine-tRNA ligase beta chain, FRSBphenylalanine tRNA ligase 1, beta, cytoplasmic, PheHB, Phenylalanine--tRNA ligase beta chain, phenylalanine-tRNA synthetase-like, beta subunit, phenylalanyl-tRNA synthetase beta chain, phenylalanyl-tRNA synthetase beta-subunit, phenylalanyl-tRNA synthetase, beta subunit, phenylalanyl-tRNA synthetase-like, beta subunit, PheRS
Host Species Mouse
Immunogen Full length human recombinant protein of human FARSB (NP_005678) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 10056
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.