Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FARSLA Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB097839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB097839 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB097839 Supplier Novus Biologicals Supplier No. NBP297761C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FARSLA Polyclonal specifically detects FARSLA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FARSLA
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias CML33pheRS, EC 6.1.1.20, FARS, FARSL, FARSLA, FRSA, PheHA, phenylalanine tRNA ligase 1, alpha, cytoplasmic, Phenylalanine--tRNA ligase alpha chain, phenylalanine-tRNA synthetase alpha-subunit, phenylalanine-tRNA synthetase-like, phenylalanine-tRNA synthetase-like, alpha subunit, phenylalanyl-tRNA synthetase alpha chain, phenylalanyl-tRNA synthetase, alpha subunit, phenylalanyl-tRNA synthetase-like, alpha subunit, PheRS
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FARSLA fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2193
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.