Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas Ligand/TNFSF6 Antibody (100419), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB176094 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176094 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176094 Supplier Novus Biologicals Supplier No. FAB126E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fas Ligand/TNFSF6 Monoclonal specifically detects Fas Ligand/TNFSF6 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry, Neutralization.

Specifications

Antigen Fas Ligand/TNFSF6
Applications ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry, Neutralization
Classification Monoclonal
Clone 100419
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Fas Ligand/TNFSF6, Pro134-Leu281, Accession # P48023
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Diabetes Research, Immunology, Inhibitors Activators and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 356
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.