Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas Ligand/TNFSF6, Mouse anti-Human, PerCP, Clone: 14C2, Novus Biologicals™
SDP

Catalog No. NB003794 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003794 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003794 Supplier Novus Biologicals Supplier No. NB10063072PCP0.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fas Ligand/TNFSF6 Monoclonal specifically detects Fas Ligand/TNFSF6 in Human samples. It is validated for ELISA, Flow Cytometry, Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen Fas Ligand/TNFSF6
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot (Negative)
Classification Monoclonal
Clone 14C2
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS
Gene Alias apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6
Gene Symbols FASLG
Host Species Mouse
Immunogen Made to Human CD178
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Diabetes Research, Immunology, Inhibitors Activators and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 356
Test Specificity NB100-63072 recognizes the human CD178 cell surface antigen, a 40kDa glycoprotein also known as Fas ligand (CD95L). CD178 is a member of the TNF family, which is expressed by activated T lymphocytes and NK cells. The protein may exist as either a membrane bound or a cleaved soluble form. CD178 plays an important role in T cell development cytotoxicity. Binding of CD178 to Fas (CD95) results in the induction of apoptosis. Clone 14C2 is not reported to have a blocking function.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.