Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas Ligand/TNFSF6 Rabbit anti-Mouse, Clone: 1, Novus Biologicals™
SDP

Catalog No. NB025222 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB025222 50 μg
NB025221 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB025222 Supplier Novus Biologicals Supplier No. NBP29073750UL

Rabbit Monoclonal Antibody

Fas Ligand/TNFSF6 Monoclonal specifically detects Fas Ligand/TNFSF6 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Fas Ligand/TNFSF6
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6
Host Species Rabbit
Immunogen Recombinant Mouse Fas Ligand/TNFSF6
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Diabetes Research, Immunology, Inhibitors Activators and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 356
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.