Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas/TNFRSF6/CD95 Antibody (028), Alexa Fluor™ 750
SDP

Catalog No. NB079038 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB079038 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB079038 Supplier Novus Biologicals Supplier No. NBP290862AF750

Rabbit Monoclonal Antibody

Fas/TNFRSF6/CD95 Monoclonal specifically detects Fas/TNFRSF6/CD95 in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Fas/TNFRSF6/CD95
Applications Flow Cytometry
Classification Monoclonal
Clone 28
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1FASTM, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Rat Fas/TNFRSF6/CD95 (NP_631933.2; Met1-Lys170).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 355
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.