Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas/TNFRSF6/CD95 Antibody (431014), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB365846 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365846 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365846 Supplier Novus Biologicals Supplier No. FAB2267X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fas/TNFRSF6/CD95 Monoclonal specifically detects Fas/TNFRSF6/CD95 in Feline samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen Fas/TNFRSF6/CD95
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 431014
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1FASTM, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant feline Fas/TNFRSF6/CD95, Ala25-Lys172, Accession # NP_001009314
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 355
Target Species Feline
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.