Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas/TNFRSF6/CD95 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP233118 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP233118 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP233118 Supplier Novus Biologicals Supplier No. NBP233118

Rabbit Polyclonal Antibody

Fas Receptor/TNFRSF6/CD95 Polyclonal specifically detects Fas Receptor/TNFRSF6/CD95 in Human, Primate samples. It is validated for Western Blot.

Specifications

Antigen Fas Receptor/TNFRSF6/CD95
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1ug/ml
Formulation 10mM PBS - BSA free with No Preservative
Gene Alias ALPS1A, Apo-1, APO-1 cell surface antigen, apoptosis antigen 1, apoptosis signaling receptor FAS, Apoptosis-mediating surface antigen FAS, APT1, CD95, CD95 antigen, Fas (TNF receptor superfamily, member 6), Fas AMA, FAS1, FASLG receptor, FASTM, mutant tum
Gene Symbols FAS
Host Species Rabbit
Immunogen Amino acids KDITSDSENSNFRNEIQSLV of human FAS (CD95) were used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 355
Test Specificity There are multiple isoforms of this protein. Researchers are encouraged to use an online tool like NCBI BLAST to determine if the antibody is suitable for their use.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.