Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas/TNFRSF6/CD95 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB003931 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003931 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003931 Supplier Novus Biologicals Supplier No. NB12013550R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fas Receptor/TNFRSF6/CD95 Polyclonal specifically detects Fas Receptor/TNFRSF6/CD95 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Fas Receptor/TNFRSF6/CD95
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ALPS1A, Apo-1, APO-1 cell surface antigen, apoptosis antigen 1, apoptosis signaling receptor FAS, Apoptosis-mediating surface antigen FAS, APT1, CD95, CD95 antigen, Fas (TNF receptor superfamily, member 6), Fas AMA, FAS1, FASLG receptor, FASTM, mutant tumor necrosis receptor superfamily member 6, TNF receptor superfamily member 6, TNFRSF6, tumor necrosis factor receptor superfamily member 6, tumor necrosis factor receptor superfamily, member 6
Gene Symbols FAS
Host Species Rabbit
Immunogen Synthetic peptide: ESLKLRRRVHETDKNC conjugated to KLH, corresponding to amino acids 29-44 of Mouse CD95.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.