Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas/TNFRSF6/CD95 Antibody (LT95), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB500503V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500503V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500503V Supplier Novus Biologicals Supplier No. NB500503V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fas Receptor/TNFRSF6/CD95 Monoclonal specifically detects Fas Receptor/TNFRSF6/CD95 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Fas Receptor/TNFRSF6/CD95
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LT95
Conjugate DyLight 405
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ALPS1A, Apo-1, APO-1 cell surface antigen, apoptosis antigen 1, apoptosis signaling receptor FAS, Apoptosis-mediating surface antigen FAS, APT1, CD95, CD95 antigen, Fas (TNF receptor superfamily, member 6), Fas AMA, FAS1, FASLG receptor, FASTM, mutant tum
Gene Symbols FAS
Host Species Mouse
Immunogen HUT-78 human T cell lymphoma cell line
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 355
Test Specificity The antibody LT95 reacts with CD95 (Fas/APO-1), a 46 kDa single chain type I glycoprotein of the tumour necrosis factor/nerve growth factor (TNF/NGF) receptor superfamily, expressed on a variety of normal and neoplastic cells. It seems that the antibody LT95 does not induce Fas mediated apoptosis, although it cross-blocks anti-Fas DX2 antibody that recognizes a functional epitope of Fas molecule.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.