Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas/TNFRSF6/CD95 Antibody (MM0041-12J3), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1106107A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1106107A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1106107A70 Supplier Novus Biologicals Supplier No. NB11061007AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fas Receptor/TNFRSF6/CD95 Monoclonal specifically detects Fas Receptor/TNFRSF6/CD95 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Fas Receptor/TNFRSF6/CD95
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0041-12J3
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ALPS1A, Apo-1, APO-1 cell surface antigen, apoptosis antigen 1, apoptosis signaling receptor FAS, Apoptosis-mediating surface antigen FAS, APT1, CD95, CD95 antigen, Fas (TNF receptor superfamily, member 6), Fas AMA, FAS1, FASLG receptor, FASTM, mutant tum
Gene Symbols FAS
Host Species Mouse
Immunogen Human recombinant Fas Receptor/TNFRSF6/CD95
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 355
Test Specificity Fas (MM0041-12J3)
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.