Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas/TNFRSF6/CD95, Human anti-Human, Clone: R-125224, Novus Biologicals™
SDP

Catalog No. NB016346 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016346 0.025 mg
NB016347 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB016346 Supplier Novus Biologicals Supplier No. NBP2811120.025MG
Add to Cart
Add to Cart

Human Monoclonal Antibody

Fas/TNFRSF6/CD95 Monoclonal specifically detects Fas/TNFRSF6/CD95 in Human samples. It is validated for Flow Cytometry, ELISA, Functional.

Specifications

Antigen Fas/TNFRSF6/CD95
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone R-125224
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA, Functional
Formulation PBS
Gene Alias Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1FASTM, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6
Gene Symbols FAS
Host Species Human
Immunogen R-125224 is generated by the humanization of the murine HFE7A anti-Fas/TNFRSF6/CD95 antibody by grafting the CDR regions to the framework regions of the human 8E10 antibody and substituting key framework residues from the murine antibody into the 8E10 sequence. The original HFE7A was derived from a hybridoma cell line generated by the fusion of NS1 myeloma cells with splenocytes from Fas-deficient mice which had been immunized with partially purified recombinant human Fas-AIC2A chimera protein consisting of the extracellular region of human Fas/TNFRSF6/CD95 antigen (aa -16 to 150) and the extracellular region of the murine IL-3 receptor AIC2 (aa 3-423). The HFE7A hybridoma was selected after screening by flow cytometry for the production of antibodies with the ability to bind to the WR19L12a transformed murine T cell lymphoma cell line expressing human Fas/TNFRSF6/CD95 or the L5178YA1 cell line expressing murine Fas/TNFRSF6/CD95, but not to the parental WR19L or L5178Y cells.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 355
Test Specificity R-125224 binds to the extracellular portion of human Fas/TNFRSF6/CD95 at an eptiope consisting of the sequence RTQNTKCRCK (aa 105-114) (pmid: 11754745). Fas is a type I membrane protein which belongs to the tumor necrosis factor (TNF) receptor/nerve growth factor (NGF) receptor superfamily. It is able to transduce apoptotic signals into the cell when bound by its ligand FasL (Fas ligand), which is primarily expressed in activated T lymphoid-myeloid lineage cells, in the eye, in reproductive organs and in some tumors. The Fas-FasL system is known to play an important role in maintaining the immune system as mice with Fas-defective lymphoproliferation (lpr) and FasL-defective generalized lymphoproliferative disease (gld) mutations develop massive lymphadenopathy and autoimmune diseases.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.