Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fascin Antibody (FSCN1/418) - IHC-Prediluted, Novus Biologicals™
SDP

Catalog No. NBP248337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP248337 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP248337 Supplier Novus Biologicals Supplier No. NBP248337
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fascin Monoclonal specifically detects Fascin in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Fascin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FSCN1/418
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias FAN1,55 kDa actin-bundling protein, fascin homolog 1, actin-bundling protein (Strongylocentrotus purpuratus), FLJ38511, HSN, p55actin bundling protein, singed (Drosophila)-like (sea urchin fascin homolog like), singed-like (fascin homolog, sea urchin), Singed-like protein, SNLfascin
Gene Symbols FSCN1
Host Species Mouse
Immunogen Full length recombinant human Fascin protein (Uniprot: Q16658)
Molecular Weight of Antigen 55 kDa
Purification Method Protein A or G purified
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 6624
Test Specificity Recognizes a protein of 55kDa, which is identified as fascin-1. Its actin binding ability is regulated by phosphorylation. Antibody to fascin-1 is a very sensitive marker for Reed-Sternberg cells and variants in nodular sclerosis, mixed cellularity, and lymphocyte depletion Hodgkins disease. It is uniformly negative in lymphoid cells, plasma cells, and myeloid cells. Fascin-1 is also expressed in dendritic cells. This marker may be helpful to distinguish between Hodgkin lymphoma and non-Hodgkin lymphoma in difficult cases. Also, the lack of expression of fascin-1 in the neoplastic follicles in follicular lymphoma may be helpful in distinguishing these lymphomas from reactive follicular hyperplasia in which the number of follicular dendritic cells is normal or increased. Antibody to fascin-1 has been suggested as a prognostic marker in neuroendocrine neoplasms of the lung as well as in ovarian cancer. Fascin-1 expression may be induced by Epstein-Barr virus (EBV) infection of B cells with the possibility that viral induction of fascin in lymphoid or other cell types must also be considered in EBV-positive cases.
Target Species Human, Rat
Content And Storage Store at 4C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.