Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fascin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP174212
Description
Fascin Polyclonal specifically detects Fascin in Mouse samples. It is validated for Western Blot.Specifications
| Fascin | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| Q61553 | |
| FSCN1 | |
| Synthetic peptides corresponding to the N terminal of Fscn1. Immunizing peptide sequence VVAHDDGRWSLQSEAHRRYFGGTEDRLSCFAQSVSPAEKWSVHIAMHPQV. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Bovine: 92%; Chicken: 85%; Zebrafish: 78%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Zebrafish | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| FAN1,55 kDa actin-bundling protein, fascin homolog 1, actin-bundling protein (Strongylocentrotus purpuratus), FLJ38511, HSN, p55actin bundling protein, singed (Drosophila)-like (sea urchin fascin homolog like), singed-like (fascin homolog, sea urchin), Singed-like protein, SNLfascin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6624 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction