Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fascin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$464.00
Specifications
| Antigen | Fascin |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Fascin Polyclonal specifically detects Fascin in Mouse samples. It is validated for Western Blot.Specifications
| Fascin | |
| Polyclonal | |
| Rabbit | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| FAN1,55 kDa actin-bundling protein, fascin homolog 1, actin-bundling protein (Strongylocentrotus purpuratus), FLJ38511, HSN, p55actin bundling protein, singed (Drosophila)-like (sea urchin fascin homolog like), singed-like (fascin homolog, sea urchin), Singed-like protein, SNLfascin | |
| FSCN1 | |
| IgG | |
| Affinity Purified |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q61553 | |
| 6624 | |
| Synthetic peptides corresponding to the N terminal of Fscn1. Immunizing peptide sequence VVAHDDGRWSLQSEAHRRYFGGTEDRLSCFAQSVSPAEKWSVHIAMHPQV. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title