Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fascin Antibody (OTI3B4), PE, Novus Biologicals™
SDP

Catalog No. NB288970 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288970 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288970 Supplier Novus Biologicals Supplier No. NBP271294PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fascin Monoclonal antibody specifically detects Fascin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Fascin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3B4
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias FAN1,55 kDa actin-bundling protein, fascin homolog 1, actin-bundling protein (Strongylocentrotus purpuratus), FLJ38511, HSN, p55actin bundling protein, singed (Drosophila)-like (sea urchin fascin homolog like), singed-like (fascin homolog, sea urchin), Singed-like protein, SNLfascin
Host Species Mouse
Immunogen Full length human recombinant protein of human FSCN1 (NP_003079) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 6624
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.