Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Fast Green FCF, Electrophoresis Reagent

Catalog No. p-7076683
Click to view available options
Quantity:
100 g

Catalog No. AAJ6657022

Supplier: Thermo Scientific Chemicals J6657022

CAS: 2353-45-9 | C37H34N2Na2O10S3, C37H34N2Na2O10S3 | 808.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2353-45-9
Molecular Formula C37H34N2Na2O10S3, C37H34N2Na2O10S3
Synonym C.I. 42053

Specifications

Content And Storage Ambient temperatures; Store under Nitrogen
Formula Weight 808.86
Melting Point ≈290°C (dec.)
InChI Key XJBPDZVCYTYRIN-UHFFFAOYSA-L
SMILES [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
IUPAC Name disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
Molecular Weight (g/mol) 808.84
Quantity 100 g
Sensitivity Hygroscopic
Form Powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.