Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Skeletal Myosin Heavy Chain 1 Antibody (LM5), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP250401AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250401AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250401AI Supplier Novus Biologicals Supplier No. NBP250401AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fast Skeletal Myosin Heavy Chain 1 Monoclonal specifically detects Fast Skeletal Myosin Heavy Chain 1 in Chicken samples. It is validated for Western Blot.

Specifications

Antigen Fast Skeletal Myosin Heavy Chain 1
Applications Western Blot
Classification Monoclonal
Clone LM5
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Purified chicken pectorial muscle
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.