Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FastAb Adjuvant

Catalog No. mp08642901 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

FastAb Adjuvant is a novel, water-soluble mouse adjuvant designed specifically for preparing monoclonal and polyclonal antibodies. It can be used as an alternative to the classic Freund's adjuvant.

FastAb Adjuvant is a novel, water-soluble mouse adjuvant designed specifically for preparing monoclonal and polyclonal antibodies. Compared to Freund's adjuvant, FastAb Adjuvant utilizes simpler preparation steps (thus shorter time), a lower dosage of immunogen, and no emulsification (and is hence capable of maintaining the native structure of the antigen). FastAb Adjuvant can be applied to monoclonal Ab screening and preparation with mouse hybridoma technology, phage display antibody library technology or single B cell cloning technology.

  • Easy to use. No emulsification - just mix with antigen and it is ready to be used for intramuscular injection.
  • Simple and short protocol. 2- or 3-weeks immunization with a total of 2 injections.
  • Reduced antigen usage, with a single dose of 0.1-50 μg.
  • Sterile, safe, and non-toxic; thus ensures good animal welfare.
  • Allows for the generation of antibodies with high titer and affinity.
  • As emulsification is not required, the native immunogen structure can be maintained. Suitable for both linear and conformational epitopes.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature
For Use With (Application) RUO
Formulation Liquid
Quantity 1 mL

For research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.