Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fasudil dihydrochloride, 99+%

Catalog No. aaj60594md
Click to view available options
Quantity:
250 mg
500 mg
1 g

CAS: 203911-27-7 | C14H19Cl2N3O2S | 364.285 g/mol

Fasudil dihydrochloride is used in the inhibition of contractile responses to potassium chloride, phenylephnne (PHE), prostaglandin and hepatic stellate cells spreading. It is also used in the formation of stress fibers and in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fasudil dihydrochloride is used in the inhibition of contractile responses to potassium chloride, phenylephnne (PHE), prostaglandin and hepatic stellate cells spreading. It is also used in the formation of stress fibers and in medicine.

Solubility
Soluble in water and dimethylsulfoxide.

Notes
Incompatible with strong oxidizing agents. Sore in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203911-27-7
Molecular Formula C14H19Cl2N3O2S
Molecular Weight (g/mol) 364.285
MDL Number MFCD00153805
InChI Key NOXXIYDYFSNHDF-UHFFFAOYSA-N
PubChem CID 16219471
IUPAC Name 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;dihydrochloride
SMILES C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl.Cl

Specifications

Color White
Quantity 250 mg
Merck Index 14,3942
Synonym fasudil dihydrochloride, ha-1077 dihydrochloride, ha 1077, dihydrochloride, 5-1,4-diazepan-1-yl sulfonyl isoquinoline dihydrochloride, 5-isoquinolinesulfonyl homopiperazine, 2hcl, 5-isoquinolinesulfonyl homopiperazine,2hcl, 1-5-isoquinolinylsulfonyl homopiperazine dihydrochloride, 1-5-isoquinoline-sulfonyl-homopiperazine, fasudil and ha 1077, dihydrochloride, ha-1077 dihydrochloride, solid hplc
Solubility Information Soluble in water and dimethylsulfoxide.
Formula Weight 364.29
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Fasudil dihydrochloride
RTECSNumber HM4031166
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.