Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fasudil hydrochloride, Tocris Bioscience™

Catalog Number 054110
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
105628-07-7
C14H18ClN3O2S
327.827
LFVPBERIVUNMGV-UHFFFAOYSA-N
fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br
163751
5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl
This item is not returnable. View return policy
105628-07-7
C14H18ClN3O2S
327.827
LFVPBERIVUNMGV-UHFFFAOYSA-N
fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br
163751
5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl

Inhibitor of cyclic nucleotide dependent- and Rho-kinases

Fasudil hydrochloride is a cyclic nucleotide-dependent protein kinase inhibitor and Rho-associated kinase inhibitor (IC50 = 10.7 μM). Fasudil suppresses MMP-2 expression and induces apoptosis in glioblastoma cells in vivo. Fasudil is a Ca2+ antagonist, vasodilator and inhibits proliferation of vascular smooth muscle cells. Fasudil binds to α-synuclein to reduce aggregate formation in cellular models of Parkinson's disease, also displays neuroprotective properties and increases survival of dopaminergic neurons in vivo.
Quantity 10 mg
Formula Weight 327.83
Percent Purity >98%
Chemical Name or Material Fasudil hydrochloride
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit