Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAT1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP23227A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP23227A70 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP23227A70 Supplier Novus Biologicals Supplier No. NBP232275AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAT1 Polyclonal specifically detects FAT1 in Human samples. It is validated for Western Blot.

Specifications

Antigen FAT1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila)
Gene Symbols FAT1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 4484 to 4534 of human FAT Tumor Suppressor Homolog 1 using the numbering given in entry NP_005236.2 (GeneID 2195).
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 2195
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.