Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAT1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP232275G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232275G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232275G Supplier Novus Biologicals Supplier No. NBP232275G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAT1 Polyclonal specifically detects FAT1 in Human samples. It is validated for Western Blot.

Specifications

Antigen FAT1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila)
Gene Symbols FAT1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 4484 to 4534 of human FAT Tumor Suppressor Homolog 1 using the numbering given in entry NP_005236.2 (GeneID 2195).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2195
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.