Learn More
Description
Specifications
Specifications
| Antigen | FAT1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila) |
| Gene Symbols | FAT1 |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 4484 to 4534 of human FAT Tumor Suppressor Homolog 1 using the numbering given in entry NP_005236.2 (GeneID 2195). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
