Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAT10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15833720UL
Description
FAT10 Polyclonal specifically detects FAT10 in Human samples. It is validated for Western Blot.Specifications
FAT10 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
O15205 | |
UBD | |
Synthetic peptides corresponding to UBD(ubiquitin D) The peptide sequence was selected from the N terminal of UBD. Peptide sequence RSKTKVPVQDQVLLLGSKILKPRRSLSSYGIDKEKTIHLTLKVVKPSDEE. | |
Affinity Purified | |
RUO | |
10537 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Diubiquitin, FAT10diubiquitin, GABBR1, UBD-3, ubiquitin D, Ubiquitin-like protein FAT10 | |
Rabbit | |
18 kDa | |
20 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction