Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAT2 Antibody (8C5), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP24371A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP24371A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP24371A30 Supplier Novus Biologicals Supplier No. NBP254371AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAT2 Monoclonal specifically detects FAT2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FAT2
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8C5
Conjugate Alexa Fluor 350
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias protocadherin Fat 2, CDHF8, CDHR9, FAT tumor suppressor homolog 2 (Drosophila), HFAT2, MEGF1
Gene Symbols FAT2
Host Species Mouse
Immunogen Purified recombinant human FAT2 protein. (Uniprot: Q9NYQ8)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2196
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.