Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAT4 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP178381A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP178381A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP178381A32 Supplier Novus Biologicals Supplier No. NBP178381AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAT4 Polyclonal specifically detects FAT4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen FAT4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cadherin family member 14, cadherin-related family member 11, CDHR11, FAT tumor suppressor homolog 4 (Drosophila), FAT-J, fat-like cadherin protein FAT-J, FLJ77878, hFat4, Nbla00548, putative protein product of Nbla00548
Gene Symbols FAT4
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human FAT4 protein (between residues 4000-4500) [UniProt Q6V0I7]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 79633
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.