Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FATE1 Antibody (OTI1A1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP272310A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272310A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272310A32 Supplier Novus Biologicals Supplier No. NBP272310AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FATE1 Monoclonal specifically detects FATE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FATE1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1A1
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Cancer/testis antigen 43, CT43BJ-HCC-2 antigen, FATEfetal and adult testis expressed transcript protein, fetal and adult testis expressed 1, fetal and adult testis-expressed transcript protein, Tumor antigen BJ-HCC-2
Gene Symbols FATE1
Host Species Mouse
Immunogen Full length recombinant protein of human FATE1 (NP_149076) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 89885
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.