Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FATP1/SLC27A1 Antibody (308420) [DyLight 594], Novus Biologicals™
SDP

Catalog No. NB372603 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372603 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372603 Supplier Novus Biologicals Supplier No. FAB3304M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FATP1/SLC27A1 Monoclonal antibody specifically detects FATP1/SLC27A1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen FATP1/SLC27A1
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 308420
Conjugate DyLight 594
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ACSVL5FATP-1, EC 6.2.1, EC 6.2.1.-, FATP1Solute carrier family 27 member 1, FATPlong-chain fatty acid transport protein 1, Fatty acid transport protein 1, FLJ00336, MGC71751, solute carrier family 27 (fatty acid transporter), member 1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human FATP1, Met1-Leu646, Accession # Q6PCB7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 376497
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.