Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FATP2/SLC27A2 Antibody (466106), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB296230 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296230 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296230 Supplier Novus Biologicals Supplier No. FAB4659U

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

FATP2/SLC27A2 Monoclonal antibody specifically detects FATP2/SLC27A2 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen FATP2/SLC27A2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 466106
Conjugate Alexa Fluor 350
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ACSVL1FATP-2, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.3, EC 6.2.1.7, FACVL1very long-chain fatty-acid-coenzyme A ligase 1, FATP2THCA-CoA ligase, Fatty acid transport protein 2, Fatty-acid-coenzyme A ligase, very long-chain 1, hFACVL1, HsT17226, Long-chain-fatty-acid--CoA ligase, solute carrier family 27 (fatty acid transporter), member 2, Solute carrier family 27 member 2, very-long-chain acyl-CoA synthetase, VLACSVery long-chain-fatty-acid-CoA ligase, VLCSvery long-chain acyl-CoA synthetase
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human FATP2, Met1-Leu620, Accession # O14975
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11001
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.