Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FATP3/SLC27A3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB060553 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB060553 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB060553 Supplier Novus Biologicals Supplier No. NBP299424AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FATP3/SLC27A3 Polyclonal specifically detects FATP3/SLC27A3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FATP3/SLC27A3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 6.2.1, EC 6.2.1.-, EC 6.2.1.7, FATP-3, FATP3ACSVL3MGC4365, Fatty acid transport protein 3, long-chain fatty acid transport protein 3, solute carrier family 27 (fatty acid transporter), member 3, Solute carrier family 27 member 3, Very long-chain acyl-CoA synthetase homolog 3, VLCS-3
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse FATP3/SLC27A3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11000
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.