Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FATP4/SLC27A4 Antibody (11A1), Novus Biologicals™
SDP

Catalog No. NB298336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298336 50 μL
NB298337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298336 Supplier Novus Biologicals Supplier No. NBP32632050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FATP4/SLC27A4 Monoclonal antibody specifically detects FATP4/SLC27A4 in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen FATP4/SLC27A4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 11A1
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias ACSVL4Solute carrier family 27 member 4, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.7, FATP-4, FATP4Fatty acid transport protein 4, IPS, long-chain fatty acid transport protein 4, solute carrier family 27 (fatty acid transporter), member 4
Host Species Rabbit
Immunogen A synthesized peptide derived from Human FATP4/SLC27A4 [UniProt Q6P1M0]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10999
Target Species Human, Mouse
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.