Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FATP4/SLC27A4 Antibody (342142), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB312835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312835 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312835 Supplier Novus Biologicals Supplier No. FAB3650PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FATP4/SLC27A4 Monoclonal specifically detects FATP4/SLC27A4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FATP4/SLC27A4
Applications Flow Cytometry
Classification Monoclonal
Clone 342142
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias ACSVL4Solute carrier family 27 member 4, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.7, FATP-4, FATP4Fatty acid transport protein 4, IPS, long-chain fatty acid transport protein 4, solute carrier family 27 (fatty acid transporter), member 4
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human FATP4, Met1-Leu643, Accession # Q6P1M0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10999
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.