Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FATP5/SLC27A5 Antibody (390917), HRP, Novus Biologicals™
SDP

Catalog No. NB348089 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348089 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348089 Supplier Novus Biologicals Supplier No. FAB3897H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FATP5/SLC27A5 Monoclonal specifically detects FATP5/SLC27A5 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen FATP5/SLC27A5
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 390917
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias ACSBBACS, ACSVL6BAL, BA-CoA ligase, Bile acid-CoA ligase, bile acyl-CoA synthetase, Cholate--CoA ligase, EC 6.2.1.7, FACVL3FATP-5, FATP5Solute carrier family 27 member 5, Fatty acid transport protein 5, Fatty-acid-coenzyme A ligase, very long-chain 3, FLJ22987, solute carrier family 27 (fatty acid transporter), member 5, VLACSRVery long-chain acyl-CoA synthetase homolog 2, VLCSH2Very long-chain acyl-CoA synthetase-related protein, VLCS-H2VLACS-related
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human FATP5, Met1-Leu690, Accession # Q9Y2P5
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10998
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.