Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FATP6/SLC27A6 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB341908 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341908 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341908 Supplier Novus Biologicals Supplier No. NBP281909PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FATP6/SLC27A6 Polyclonal antibody specifically detects FATP6/SLC27A6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen FATP6/SLC27A6
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias ACSVL2VLCSH1, DKFZp779M0564, EC 6.2.1, EC 6.2.1.7, FACVL2hVLCS-H1, FATP1, FATP6, FATP-6, solute carrier family 27 (fatty acid transporter), member 6, Solute carrier family 27 member 6, very long-chain 2, Very long-chain acyl-CoA synthetase homolog 1, VLCS-H1
Host Species Rabbit
Immunogen SLC27A6 antibody was raised against a 16 amino acid peptide near the amino terminus of human SLC27A6. The immunogen is located within amino acids 180 - 230 of SLC27A6. Amino Acid Squence: KEKLSTSPDEPVPRSH
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 28965
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.