Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fatty Acid Synthase/FASN Antibody (325), Novus Biologicals™
SDP

Catalog No. NBP243822 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243822 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243822 Supplier Novus Biologicals Supplier No. NBP243822
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot

Fatty Acid Synthase/FASN Monoclonal specifically detects Fatty Acid Synthase/FASN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Fatty Acid Synthase/FASN
Applications Western Blot
Classification Monoclonal
Clone 325
Conjugate Unconjugated
Dilution Western Blot 1:500-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS with No Preservative
Gene Alias EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1
Gene Symbols FASN
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human Fatty Acid Synthase. The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Diabetes Research, Lipid and Metabolism, Neuroendocrinology
Primary or Secondary Primary
Gene ID (Entrez) 2194
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.