Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fatty Acid Synthase/FASN Antibody, Novus Biologicals™
SDP

Catalog No. NB400114 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400114 0.1 mL
NB400114SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400114 Supplier Novus Biologicals Supplier No. NB400114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 23 publications

Fatty Acid Synthase/FASN Polyclonal specifically detects Fatty Acid Synthase/FASN in Human, Mouse, Rat, Porcine, Chicken, Feline, Hamster, Plant, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen Fatty Acid Synthase/FASN
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:2000, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500immunoprecipitation 1:100, Knockdown Validated
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. P19096
Gene Alias EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1
Gene Symbols FASN
Host Species Rabbit
Immunogen A synthetic peptide, conjugated to KLH, made near the N-terminus of mouse FAS. [Swiss-Prot# P19096]
Molecular Weight of Antigen 272 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Lipid and Metabolism, Neuroendocrinology
Primary or Secondary Primary
Gene ID (Entrez) 2194
Target Species Human, Mouse, Rat, Pig, Chicken, Feline, Hamster, Plant, Primate
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.