Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBG4 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB345145 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345145 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345145 Supplier Novus Biologicals Supplier No. NB100466PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FBG4 Polyclonal specifically detects FBG4 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen FBG4
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias FBG4FBXO26, F-box only protein 17, F-box protein 17, F-box protein FBG4, Fbx17, FBX26F-box only protein 26, FLJ11798, FLJ25205, FLJ58300, MGC9379
Host Species Rabbit
Immunogen A synthetic peptide contianing residues 70-85 (LQLARDRSAEGRALYA) of the human FBG4 protein. There is 100% sequence identity between the immunogen and mouse and rat sequences.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115290
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.