Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fbx32 Antibody, Novus Biologicals™
SDP

Catalog No. NB387838 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387838 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387838 Supplier Novus Biologicals Supplier No. NBP323417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fbx32 Polyclonal antibody specifically detects Fbx32 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Fbx32
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias atrogin 1, atrogin-1, ATROGIN1, F-box only protein 32, F-box protein 32, Fbx32, FLJ32424, MAFBX, MAFbxMGC33610, Muscle atrophy F-box protein
Host Species Rabbit
Immunogen A synthetic peptide (coupled to carrier protein) corresponding to amino acids from the N-terminal region of mouse Fbx32. This sequence is highly homologous to human and rat Fbx32 (F-box only protein 32). [UniProt# Q9CPU7]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cell Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 114907
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.