Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXL16 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen FBXL16
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP157613
SDP
View Documents
Novus Biologicals
NBP157613
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

FBXL16 Polyclonal specifically detects FBXL16 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FBXL16
Polyclonal
Rabbit
Q8N461
146330
Synthetic peptides corresponding to FBXL16(F-box and leucine-rich repeat protein 16) The peptide sequence was selected from the middle region of FBXL16. Peptide sequence GCPLLTTTGLSGLVQLQELEELELTNCPGATPELFKYFSQHLPRCLVIE.
Primary
Western Blot
Unconjugated
RUO
C16orf22, c380A1.1, c380A1.1 (novel protein), chromosome 16 open reading frame 22, FBL16, F-box and leucine-rich repeat protein 16Fbl16, F-box/LRR-repeat protein 16, FLJ33735, MGC33974
FBXL16
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.