Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXL16 Antibody, Novus Biologicals™
SDP

Catalog No. NBP238789 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP238789 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP238789 Supplier Novus Biologicals Supplier No. NBP238789
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FBXL16 Polyclonal specifically detects FBXL16 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FBXL16
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q8N461
Gene Alias C16orf22, c380A1.1, c380A1.1 (novel protein), chromosome 16 open reading frame 22, FBL16, F-box and leucine-rich repeat protein 16Fbl16, F-box/LRR-repeat protein 16, FLJ33735, MGC33974
Gene Symbols FBXL16
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: CPRITDMALEYVACDLHRLEELVLDRCVRITDTGLSYLSTMSSLRSLYLRWCCQVQDFGLKH
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146330
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.