Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXL2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15527920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15527920 20 μL
NBP155279 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15527920 Supplier Novus Biologicals Supplier No. NBP15527920UL

Rabbit Polyclonal Antibody

FBXL2 Polyclonal specifically detects FBXL2 in Human samples. It is validated for Western Blot.

Specifications

Antigen FBXL2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q9UKC9
Gene Alias FBL2FBL3F-box protein containing leucine-rich repeats, F-box and leucine-rich repeat protein 2DKFZp564P0622, F-box protein FBL2/FBL3, F-box/LRR-repeat protein 2
Gene Symbols FBXL2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to FBXL2(F-box and leucine-rich repeat protein 2) The peptide sequence was selected from the N terminal of FBXL2. Peptide sequence NISKRCGGFLRKLSLRGCIGVGDSSLKTFAQNCRNIEHLNLNGCTKITDS.
Molecular Weight of Antigen 47 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25827
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.