Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15504620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15504620 20 μL
NBP155046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15504620 Supplier Novus Biologicals Supplier No. NBP15504620UL

Rabbit Polyclonal Antibody

FBXO3 Polyclonal specifically detects FBXO3 in Human samples. It is validated for Western Blot.

Specifications

Antigen FBXO3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q9UK99
Gene Alias DKFZp564B092, F-box protein 3, F-box protein FBX3, FBX3FBAF-box only protein 3
Gene Symbols FBXO3
Host Species Rabbit
Immunogen Synthetic peptides corresponding to FBXO3(F-box protein 3) The peptide sequence was selected from the N terminal of FBXO3. Peptide sequence NCCYVSRRLSQLSSHDPLWRRHCKKYWLISEEEKTQKNQCWKSLFIDTYS.
Molecular Weight of Antigen 47 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26273
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.