Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | FBXO3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15504620
![]() |
Novus Biologicals
NBP15504620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155046
![]() |
Novus Biologicals
NBP155046 |
100 μL |
Each for $487.50
|
|
|||||
Description
FBXO3 Polyclonal specifically detects FBXO3 in Human samples. It is validated for Western Blot.Specifications
FBXO3 | |
Polyclonal | |
Rabbit | |
Q9UK99 | |
26273 | |
Synthetic peptides corresponding to FBXO3(F-box protein 3) The peptide sequence was selected from the N terminal of FBXO3. Peptide sequence NCCYVSRRLSQLSSHDPLWRRHCKKYWLISEEEKTQKNQCWKSLFIDTYS. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DKFZp564B092, F-box protein 3, F-box protein FBX3, FBX3FBAF-box only protein 3 | |
FBXO3 | |
IgG | |
47 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title