Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO31 Antibody (OTI1H1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N272208A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272208A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272208A700 Supplier Novus Biologicals Supplier No. NBP272208AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FBXO31 Monoclonal specifically detects FBXO31 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FBXO31
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1H1
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp434B027, F-box only protein 31, F-box protein 31, FBX14DKFZp434J1815, Fbx31, FBXO14FLJ22477, MGC15419, MGC9527, pp2386, putative breast cancer tumor-suppressor, SCF ubiquitin ligase specificity factor
Gene Symbols FBXO31
Host Species Mouse
Immunogen Full length recombinant protein of human FBXO31 (NP_079011) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 79791.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.